Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGC95210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005532.3 | NP_001005532.2 | 4 | 2069 | ||
XM_006247676.2 | XP_006247738.1 | 3 | 1700 | |||
XM_008768322.1 | XP_008766544.1 | 3 | 1700 | |||
XM_008768323.1 | XP_008766545.1 | 3 | 1570 | |||
XM_008768324.1 | XP_008766546.1 | 3 | 1572 | |||
XM_008768325.1 | XP_008766547.1 | 4 | 1644 | |||
XM_008768326.1 | XP_008766548.1 | 4 | 1700 | |||
XM_008768327.1 | XP_008766549.1 | 3 | 1484 | |||
XM_008768328.1 | XP_008766550.1 | 3 | 1700 | |||
XM_008768329.1 | XP_008766551.1 | 3 | 1700 | |||
XM_008768330.1 | XP_008766552.1 | 3 | 2781 | |||
XM_008768331.1 | XP_008766553.1 | 1 | 1406 | |||
GenBank mRNA | BC082057.1 | - | 3 | 1430 | 7132329 | |
BC088445.1 | - | 3 | 1689 | 7126436 | ||
BC099187.1 | - | 4 | 2069 | 7383366 |