Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbfox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134498.2 | NP_001127970.1 | 4 | 475 | ||
XM_008768343.1 | XP_008766565.1 | 4 | 2455 | |||
XM_008768344.1 | XP_008766566.1 | 4 | 2454 | |||
XM_008768345.1 | XP_008766567.1 | 4 | 2453 | |||
XM_008768346.1 | XP_008766568.1 | 4 | 2452 |