Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mzt2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105860.1 | NP_001099330.1 | 2 | 695 | ||
XM_006248729.1 | XP_006248791.1 | 3 | 327 | |||
XM_006248730.2 | XP_006248792.1 | 3 | 767 | |||
XM_006248731.2 | XP_006248793.1 | 3 | 761 |