Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105867.1 | NP_001099337.1 | 6 | 1535 | ||
XM_006248594.1 | XP_006248656.1 | 6 | 1306 |