Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem30c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105889.1 | NP_001099359.1 | 4, 5 | 511 | ||
XM_008768632.1 | XP_008766854.1 | 4, 5 | 748 |