Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006955.1 | NP_001006956.1 | 4 | 531 | ||
XM_006248204.2 | XP_006248266.1 | 5 | 723 | |||
XM_006248205.2 | XP_006248267.1 | 4 | 672 | |||
XM_006248206.2 | XP_006248268.1 | 5 | 574 | |||
XM_008768633.1 | XP_008766855.1 | 4 | 514 | |||
GenBank mRNA | BC083649.1 | - | 4 | 531 | 7135100 |