Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chodl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105894.1 | NP_001099364.1 | 1 | 447 | ||
XM_006248003.2 | XP_006248065.1 | 1 | 501 | |||
XM_006248004.2 | XP_006248066.1 | 1 | 429 |