Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wipi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007615.1 | NP_001007616.1 | 4 | 576 | ||
GenBank mRNA | BC079184.1 | - | 4 | 576 | 7131054 |