Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbed5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105924.1 | NP_001099394.1 | 4 | 1760 | ||
XM_006249235.2 | XP_006249297.1 | 3 | 1722 |