Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105932.1 | NP_001099402.1 | 3 | 300 | ||
XM_006249362.2 | XP_006249424.1 | 4 | 326 | |||
GenBank mRNA | AB578912.1 | - | 2 | 50 | ||
BC167055.1 | - | 4 | 287 | 7302374 | ||
FQ224287.1 | - | 4 | 308 |