Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100943.1 | NP_001094413.1 | 1 | 561 | ||
NM_001114392.1 | NP_001107864.1 | 1 | 761 |
Set Membership: |
G-protein coupled receptor |