Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105952.1 | NP_001099422.1 | 4 | 716 | ||
XM_006249762.2 | XP_006249824.1 | 3 | 377 |