Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181367.2 | NP_852032.1 | 4 | 335 | ||
XM_006249914.2 | XP_006249976.1 | 2 | 827 | |||
XM_006249915.2 | XP_006249977.1 | 2 | 832 | |||
XM_006249916.2 | XP_006249978.1 | 5 | 897 | |||
XM_006249917.1 | XP_006249979.1 | 2 | 188 | |||
XM_006249918.2 | XP_006249980.1 | 2 | 822 | |||
XM_006249919.2 | XP_006249981.1 | 5 | 897 | |||
XM_008769545.1 | XP_008767767.1 | 4 | 423 | |||
XM_008769546.1 | XP_008767768.1 | 2 | 822 | |||
GenBank mRNA | AF527619.1 | - | 2 | 143 | ||
AY273890.1 | - | 4 | 335 | |||
AY724502.1 | - | 4 | 335 | |||
BC128722.1 | - | 5 | 411 | 8362986 |