Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Swt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105960.1 | NP_001099430.1 | 8 | 1320 | ||
XM_008769613.1 | XP_008767835.1 | 8 | 1349 |