Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082580.1 | NP_001076049.1 | 6 | 777 | ||
NM_182668.2 | NP_872609.2 | 6 | 745 | |||
GenBank mRNA | AY300752.1 | - | 5 | 608 | ||
BC087146.1 | - | 6 | 726 | 7129801 | ||
FQ213884.1 | - | 528 | ||||
FQ214471.1 | - | 775 | ||||
FQ227281.1 | - | 774 |