Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082580.1 | NP_001076049.1 | 4 | 519 | ||
NM_182668.2 | NP_872609.2 | 4 | 487 | |||
GenBank mRNA | AY300752.1 | - | 3 | 350 | ||
BC087146.1 | - | 4 | 468 | 7129801 | ||
CB576182.1 | - | 4 | 582 | |||
CO558072.1 | - | 4 | 525 | |||
FQ214471.1 | - | 517 | ||||
FQ227281.1 | - | 516 |