Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eprs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024238.1 | NP_001019409.1 | 8 | 727 | ||
XM_006250425.2 | XP_006250487.1 | 7 | 827 | |||
XM_006250426.2 | XP_006250488.1 | 7 | 827 | |||
XM_006250427.2 | XP_006250489.1 | 3 | 207 | |||
GenBank mRNA | AY383705.1 | - | 8 | 727 |