Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105999.1 | NP_001099469.1 | 30 | 7853 | ||
XM_006250763.2 | XP_006250825.1 | 33 | 7888 | |||
XM_006250764.2 | XP_006250826.1 | 33 | 7885 | |||
XM_006250765.2 | XP_006250827.1 | 33 | 7834 | |||
XM_006250766.2 | XP_006250828.1 | 33 | 8470 | |||
XM_006250767.2 | XP_006250829.1 | 33 | 8770 | |||
XM_006250768.2 | XP_006250830.1 | 32 | 7060 | |||
XM_006250769.2 | XP_006250831.1 | 32 | 7057 | |||
XM_006250770.2 | XP_006250832.1 | 32 | 7006 | |||
XM_008770011.1 | XP_008768233.1 | 33 | 8467 |