Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106002.1 | NP_001099472.1 | 21 | 3019 | ||
GenBank mRNA | FQ213437.1 | - | 3 | 294 |