Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004211.1 | NP_001004211.1 | 5, 6 | 1002 | ||
XM_006251437.2 | XP_006251499.1 | 6, 7 | 937 | |||
GenBank mRNA | BC078919.1 | - | 5, 6 | 1002 | 7096512 | |
FQ212430.1 | - | 923 |