Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106031.1 | NP_001099501.1 | 4 | 384 | ||
XM_006251857.2 | XP_006251919.1 | 4 | 414 |