Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102549712 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252224.2 | XP_006252286.1 | 887 |
Gene Symbol: Ints9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770801.1 | XP_008769023.1 | 10 | 1276 |