Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcbtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199084.1 | NP_954515.1 | 15 | 1861 | ||
XM_008770845.1 | XP_008769067.1 | 15 | 2000 | |||
XM_008770846.1 | XP_008769068.1 | 15 | 2082 | |||
XM_008770847.1 | XP_008769069.1 | 15 | 2045 | |||
XM_008770848.1 | XP_008769070.1 | 14 | 1774 | |||
XM_008770849.1 | XP_008769071.1 | 14 | 1917 | |||
XM_008770850.1 | XP_008769072.1 | 14 | 1999 | |||
XM_008770851.1 | XP_008769073.1 | 16 | 1971 | |||
XM_008770852.1 | XP_008769074.1 | 16 | 2968 | |||
XM_008770853.1 | XP_008769075.1 | 16 | 2185 | |||
XM_008770854.1 | XP_008769076.1 | 16 | 1953 | |||
XM_008770855.1 | XP_008769077.1 | 17 | 2349 | |||
XM_008770856.1 | XP_008769078.1 | 14 | 1935 | |||
XM_008770857.1 | XP_008769079.1 | 13 | 1886 | |||
XM_008770858.1 | XP_008769080.1 | 13 | 1841 | |||
XM_008770859.1 | XP_008769081.1 | 12 | 1679 | |||
XM_008770860.1 | XP_008769082.1 | 12 | 1612 | |||
XM_008770861.1 | XP_008769083.1 | 13 | 1754 | |||
GenBank mRNA | BC061766.1 | - | 15 | 1861 | 5621190 | |
FQ211316.1 | - | 13 | 1719 | |||
FQ211373.1 | - | 13 | 1719 |