Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771053.1 | XP_008769275.1 | 29 | 3538 | ||
XM_008771054.1 | XP_008769276.1 | 29 | 3538 | |||
XM_008771055.1 | XP_008769277.1 | 30 | 3370 |