Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map1s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106070.1 | NP_001099540.1 | 6, 7 | 2794 | ||
XM_008771085.1 | XP_008769307.1 | 6, 7 | 3133 | |||
XM_008771086.1 | XP_008769308.1 | 7, 8 | 2879 | |||
GenBank mRNA | BC169058.1 | - | 6, 7 | 2766 | 7932017 | |
FQ211658.1 | - | 6, 7 | 2803 |