Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1311747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169105.1 | NP_001162576.1 | 3 | 282 | ||
GenBank mRNA | DY557818.1 | - | 3 | 282 |