Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047849.1 | NP_001041314.1 | 8, 9 | 1837 | ||
GenBank mRNA | BC082092.1 | - | 5, 6 | 683 | ||
BC161809.1 | - | 7, 8 | 1123 | 8374387 | ||
DQ480750.1 | - | 8, 9 | 1837 |