Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031642.1 | NP_001026812.1 | 4 | 539 | ||
GenBank mRNA | BC098686.1 | - | 4 | 539 | 7385870 |