Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hecw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106117.1 | NP_001099587.1 | 5 | 710 | ||
XM_008771697.1 | XP_008769919.1 | 5 | 628 |