Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024129.1 | NP_077043.1 | 2 | 93 | ||
XM_006241285.1 | XP_006241347.1 | 2 | 278 | |||
GenBank mRNA | BC083750.1 | - | 2 | 93 | 7192499 | |
FQ210569.1 | - | 2 | 108 | |||
FQ213980.1 | - | 2 | 212 | |||
FQ219808.1 | - | 2 | 126 | |||
FQ220160.1 | - | 2 | 117 | |||
FQ220209.1 | - | 2 | 159 | |||
FQ222189.1 | - | 2 | 162 | |||
FQ223184.1 | - | 2 | 167 | |||
FQ228702.1 | - | 2 | 117 | |||
FQ229041.1 | - | 2 | 181 | |||
FQ229786.1 | - | 2 | 162 | |||
Z12298.1 | - | 2 | 222 |