Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024129.1 | NP_077043.1 | 2 | 144 | ||
XM_006241285.1 | XP_006241347.1 | 2 | 329 | |||
GenBank mRNA | BC083750.1 | - | 2 | 144 | 7192499 | |
FQ210569.1 | - | 2 | 159 | |||
FQ213980.1 | - | 2 | 263 | |||
FQ219808.1 | - | 2 | 177 | |||
FQ220160.1 | - | 2 | 168 | |||
FQ220209.1 | - | 2 | 210 | |||
FQ223184.1 | - | 2 | 218 | |||
FQ228702.1 | - | 2 | 168 | |||
FQ229041.1 | - | 2 | 232 | |||
FQ229786.1 | - | 2 | 213 | |||
X59859.1 | - | 1 | 64 | |||
Z12298.1 | - | 2 | 273 |