Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017124.1 | NP_058820.1 | 5 | 415 | ||
XM_006228981.2 | XP_006229043.1 | 6 | 608 | |||
XM_008759326.1 | XP_008757548.1 | 5 | 454 | |||
GenBank mRNA | BC059144.1 | - | 5 | 456 | 6887407 | |
FQ230385.1 | - | 5 | 433 | |||
X53517.1 | - | 5 | 415 |