Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255791.2 | XP_006255853.1 | 18 | 2268 | ||
XM_006255792.2 | XP_006255854.1 | 18 | 2268 | |||
XM_006255793.2 | XP_006255855.1 | 17 | 2196 | |||
XM_214707.8 | XP_214707.6 | 18 | 2268 | |||
GenBank mRNA | FQ215148.1 | - | 18 | 2034 |