Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgbd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106198.1 | NP_001099668.1 | 5 | 1148 | ||
XM_006255807.2 | XP_006255869.1 | 5 | 1235 |