Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019167.1 | NP_062040.1 | 9 | 1421 | ||
XM_008760092.1 | XP_008758314.1 | 8 | 1653 | |||
XM_008760093.1 | XP_008758315.1 | 8 | 1653 | |||
XM_008760094.1 | XP_008758316.1 | 9 | 1150 | |||
GenBank mRNA | AB001347.1 | - | 9 | 1421 | ||
AB008551.1 | - | 8 | 1030 | |||
AF225960.1 | - | 8 | 1030 |