Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031566.1 | NP_113754.1 | 2 | 381 | ||
XM_006238335.2 | XP_006238397.1 | 2 | 1046 | |||
XM_006238336.2 | XP_006238398.1 | 2 | 1046 | |||
XM_006238339.2 | XP_006238401.1 | 2 | 1044 | |||
XM_008763762.1 | XP_008761984.1 | 2 | 788 | |||
GenBank mRNA | AB012230.2 | - | 2 | 381 | ||
AB012231.2 | - | 2 | 381 | |||
AB012232.2 | - | 2 | 381 | |||
AF112457.1 | - | 1 | 326 |