Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Penk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017139.1 | NP_058835.1 | 2 | 1067 | ||
XM_006237835.1 | XP_006237897.1 | 3 | 920 | |||
GenBank mRNA | BC083563.1 | - | 2 | 1058 | 7112833 | |
M28263.1 | - | 3 | 788 | |||
S49491.1 | - | 3 | 858 | |||
Y07503.1 | - | 2 | 1067 |