Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022924.2 | NP_075213.2 | 14 | 1845 | ||
GenBank mRNA | CO575133.1 | - | 2 | 161 | ||
FQ209979.1 | - | 4 | 518 | |||
M81397.1 | - | 5 | 684 | |||
X52835.1 | - | 14 | 1910 |