Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106220.1 | NP_001099690.1 | 2 | 1030 | ||
XM_008758899.1 | XP_008757121.1 | 3 | 1119 |