Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_218239.5 | XP_218239.5 | 4 | 853 |
Gene Symbol: LOC100912263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759488.1 | XP_008757710.1 | 1 | 129 |