Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oprl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031569.3 | NP_113757.1 | 4 | 1265 | ||
XM_006235736.2 | XP_006235798.1 | 5 | 1355 | |||
XM_006235738.2 | XP_006235800.1 | 4 | 1113 | |||
XM_006235739.2 | XP_006235801.1 | 6 | 1389 | |||
XM_008762474.1 | XP_008760696.1 | 3 | 875 | |||
GenBank mRNA | AF115267.1 | - | 4 | 1086 | ||
AY152731.1 | - | 3 | 1024 | |||
D16438.1 | - | 4 | 1076 | |||
L28144.1 | - | 4 | 1196 | |||
L29419.1 | - | 3 | 1159 | |||
L33916.1 | - | 4 | 1130 | |||
U01913.1 | - | 4 | 1205 | |||
U05239.1 | - | 4 | 1207 | |||
U07871.1 | - | 3 | 1055 |
Set Membership: |
G-protein coupled receptor |