Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106257.1 | NP_001099727.1 | 14 | 1620 | ||
XM_006229002.2 | XP_006229064.1 | 14 | 1582 |