Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229185.1 | XP_006229247.1 | 5 | 2555 | ||
XM_006229186.1 | XP_006229248.1 | 5 | 2173 | |||
XM_006229187.1 | XP_006229249.1 | 5 | 2126 | |||
XM_006229188.2 | XP_006229250.1 | 5 | 2054 | |||
XM_008759453.1 | XP_008757675.1 | 4 | 2378 | |||
XM_008759454.1 | XP_008757676.1 | 4 | 1922 |