Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047852.2 | NP_001041317.2 | 4 | 517 | ||
XM_006227652.2 | XP_006227714.1 | 5 | 684 | |||
XM_006227653.2 | XP_006227715.1 | 3 | 513 | |||
XM_006227654.2 | XP_006227716.1 | 4 | 541 | |||
XM_006227655.2 | XP_006227717.1 | 4 | 547 | |||
XM_006227656.2 | XP_006227718.1 | 4 | 541 | |||
XM_008758605.1 | XP_008756827.1 | 4 | 1676 | |||
GenBank mRNA | BC161968.1 | - | 4 | 517 | 9028507 |