Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134563.1 | NP_001128035.1 | 1, 2 | 1681 | ||
XM_008759507.1 | XP_008757729.1 | 2, 3 | 3269 | |||
XM_008759508.1 | XP_008757730.1 | 2, 3 | 3074 |