Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Art2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198735.2 | NP_942030.1 | 2 | 333 | ||
XM_008759694.1 | XP_008757916.1 | 2 | 561 | |||
XM_008759695.1 | XP_008757917.1 | 2 | 561 | |||
XM_008759696.1 | XP_008757918.1 | 2 | 1182 | |||
XM_008759697.1 | XP_008757919.1 | 3 | 1550 | |||
XM_008759698.1 | XP_008757920.1 | 5 | 922 | |||
XM_008759699.1 | XP_008757921.1 | 2 | 408 | |||
XM_008759700.1 | XP_008757922.1 | 4 | 653 | |||
XM_008759701.1 | XP_008757923.1 | 2 | 476 | |||
XM_008759702.1 | XP_008757924.1 | 2 | 1182 | |||
GenBank mRNA | BC099070.1 | - | 2 | 333 | 7366134 | |
FQ227811.1 | - | 1 | 106 | |||
FQ230306.1 | - | 4 | 669 | |||
M85193.1 | - | 2 | 310 | |||
X99122.1 | - | 5 | 927 | |||
X99123.1 | - | 3 | 383 | |||
X99124.1 | - | 3 | 400 |