Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubfd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034911.1 | NP_001030083.1 | 6 | 813 | ||
XM_006230189.2 | XP_006230251.1 | 7 | 1157 | |||
GenBank mRNA | BC101866.1 | - | 6 | 813 | 7938969 | |
CK839597.1 | - | 2 | 419 |