Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asphd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759931.1 | XP_008758153.1 | 1 | 1539 | ||
XM_215083.7 | XP_215083.2 | 1 | 1539 |