Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam175b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106307.1 | NP_001099777.2 | 2 | 125 | ||
GenBank mRNA | FQ218234.1 | - | 2 | 114 |