Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019195.2 | NP_062068.1 | 4 | 628 | ||
XM_006248262.2 | XP_006248324.1 | 5 | 1321 | |||
XM_006248263.2 | XP_006248325.1 | 5 | 1321 | |||
XM_006248264.2 | XP_006248326.1 | 5 | 1321 | |||
XM_006248265.2 | XP_006248327.1 | 5 | 1321 | |||
XM_006248266.2 | XP_006248328.1 | 4 | 650 | |||
XM_006248267.2 | XP_006248329.1 | 4 | 1207 | |||
XM_006248268.2 | XP_006248330.1 | 5 | 1519 | |||
GenBank mRNA | AF017437.1 | - | 4 | 514 | ||
BC085740.1 | - | 4 | 628 | 7189894 | ||
D87659.1 | - | 4 | 538 | |||
FQ209533.1 | - | 3 | 251 | |||
FQ232318.1 | - | 3 | 251 |